2-(Bromomethyl)aniline, N-BOC protected ≥96%

Supplier: Ambeed
Danger

A657979-1G A657979-250MG A657979-100MG A657979-5G
77614-030EA 132.77 USD
77614-030 77673-790 77884-535 77884-536
2-(Bromomethyl)aniline, N-BOC protected ≥96%
2-(Bromomethyl)aniline, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂
MDL Number: MFCD11616720
CAS Number: 166329-43-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR