2-(2-Iodoethyl)-1,3-dioxolane ≥96%

Supplier: Ambeed
Warning

A657707-10G A657707-1G
77893-322EA 186 USD
77893-322 77893-323
2-(2-Iodoethyl)-1,3-dioxolane ≥96%

Precaución: for Research Use Only

Formula: C₅H₉IO₂
MDL Number: MFCD25964997
CAS Number: 83665-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR