(3S,4S)-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A657539-1G A657539-250MG
77884-530EA 102 USD
77884-530 77884-531
(3S,4S)-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate ≥95%
(3S,4S)-tert-Butyl 4-amino-3-fluoropiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₉FN₂O₂
MDL Number: MFCD26516106
CAS Number: 1228185-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR