1-(1-Trityl-1H-imidazol-4-yl)ethanone ≥95%
Supplier: Ambeed
|
Warning
|
A657405-1G
A657405-250MG
A657405-100MG
77602-678EA
45.1
USD
77602-678
77669-794
77873-058
1-(1-Trityl-1H-imidazol-4-yl)ethanone ≥95%
1-(1-Trityl-1H-imidazol-4-yl)ethanone
Precaución: for Research Use Only
Formula:
C₂₄H₂₀N₂O |
MDL Number:
MFCD17215519 CAS Number: 116795-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...