trans-4-((Dimethylamino)methyl)cyclohexanamine dihydrochloride ≥98%

Supplier: Ambeed
Warning

A657284-100MG A657284-1G A657284-250MG
77783-605EA 21 USD
77783-605 77783-606 77783-607
trans-4-((Dimethylamino)methyl)cyclohexanamine dihydrochloride ≥98%
trans-4-((Dimethylamino)methyl)cyclohexanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₂Cl₂N₂
MDL Number: MFCD30802603
CAS Number: 1356954-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR