HOOCCH2CH2O-PEG7-CH2CH2COOH ≥95%

Supplier: Ambeed
Warning

Synonyms: Bis-PEG8-acid

A656951-1G A656951-5G A656951-100MG A656951-250MG
77598-666EA 199.88 USD
77598-666 77598-668 77622-428 77622-430
HOOCCH2CH2O-PEG7-CH2CH2COOH ≥95%
HOOCCH2CH2O-PEG7-CH2CH2COOH

Precaución: for Research Use Only

Formula: C₂₀H₃₈O₁₂
MW: 470.5085 g/mol
MDL Number: MFCD29917781
CAS Number: 1246189-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR