(R)-(-)-N-Benzyl-2-phenylglycinol 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-2-(Benzylamino)-2-phenylethanol

A656783-250MG A656783-1G A656783-100MG
77617-750EA 8.24 USD
77617-750 77747-342 77747-341
(R)-(-)-N-Benzyl-2-phenylglycinol 98+%
(R)-(-)-N-Benzyl-2-phenylglycinol

Precaución: for Research Use Only

Formula: C₁₅H₁₇NO
MDL Number: MFCD00674035
CAS Number: 14231-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR