tert-Butyl (8-hydroxyoctyl)carbamate ≥95%

Supplier: Ambeed
Warning

A656709-5G A656709-25G
77595-002EA 121.98 USD
77595-002 77747-340
tert-Butyl (8-hydroxyoctyl)carbamate ≥95%
tert-Butyl (8-hydroxyoctyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₇NO₃
MW: 245.35 g/mol
MDL Number: MFCD02094528
CAS Number: 144183-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR