5-Bromo-2-methylphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (5-Bromo-2-methylphenyl)boronic acid

A656646-100MG A656646-250MG
77884-528EA 50 USD
77884-528 77884-529
5-Bromo-2-methylphenylboronic acid ≥98%
5-Bromo-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MDL Number: MFCD16295061
CAS Number: 774608-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR