3-(3-Hydroxypropyl)benzeneboronic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: (3-(3-Hydroxypropyl)phenyl)boronic acid

A655931-250MG A655931-1G A655931-5G
77627-314EA 122.57 USD
77627-314 77878-481 77878-482
3-(3-Hydroxypropyl)benzeneboronic acid ≥98%
3-(3-Hydroxypropyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₃
MDL Number: MFCD04115650
CAS Number: 736989-98-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR