Methyl (1S,4aS,7S,7aR)-7-methyl-5-oxo-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate ≥98%

Supplier: Ambeed
Warning

Synonyms: Verbenalin

A655719-1MG A655719-10MG
77602-676EA 29.73 USD
77602-676 77669-786
Methyl (1S,4aS,7S,7aR)-7-methyl-5-oxo-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate ≥98%
Methyl (1S,4aS,7S,7aR)-7-methyl-5-oxo-1-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1,4a,5,6,7,7a-hexahydrocyclopenta[c]pyran-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₄O₁₀
MW: 388.37 g/mol
MDL Number: MFCD27996262
CAS Number: 548-37-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR