Methyl trans,trans-9,12-octadecadienoate 98+%

Supplier: Ambeed
Warning

Synonyms: Methyl trans,trans-9,12-Octadecadienoate, Methyl Linolelaidate, (9E,12E)-Methyl octadeca-9,12-dienoate, trans,trans-9,12-Octadecadienoic Acid Methyl Ester, Linolelaidic Acid Methyl Ester

A655652-250MG A655652-1G
77602-674EA 50.23 USD
77602-674 77669-784
Methyl trans,trans-9,12-octadecadienoate 98+%
Methyl trans,trans-9,12-octadecadienoate

Precaución: for Research Use Only

Formula: C₁₉H₃₄O₂
MDL Number: MFCD00069997
CAS Number: 2566-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR