3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester ≥96%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)morpholine, 3-Fluoro-2-(morpholin-4-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 4-[3-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]morpholine

A655564-100MG A655564-1G A655564-250MG
77901-057EA 24 USD
77901-057 77901-562 77901-825
3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester ≥96%
3-Fluoro-2-(morpholin-4-yl)pyridine-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BFN₂O₃
MDL Number: MFCD07781241
CAS Number: 957198-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR