5-(Pyridin-4-yl)isophthalic acid ≥97%

Supplier: Ambeed
Warning

A654652-1G A654652-250MG
77747-336EA 133 USD
77747-336 77747-337
5-(Pyridin-4-yl)isophthalic acid ≥97%
5-(Pyridin-4-yl)isophthalic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₄
MDL Number: MFCD30342484
CAS Number: 1262962-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR