7-Azaspiro[3.5]nonan-2-ol hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: 7-azaspiro[3.5]nonan-2-ol hydrochloride

A654528-250MG A654528-1G A654528-5G
77614-022EA 97.85 USD
77614-022 77684-546 77684-547
7-Azaspiro[3.5]nonan-2-ol hydrochloride ≥98%
7-Azaspiro[3.5]nonan-2-ol hydrochloride

Precaución: For research use only.

Formula: C₈H₁₅NO·ClH
MW: 177.672 g/mol
MDL Number: MFCD20486137
CAS Number: 587869-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR