Ethyl-4-(bromomethyl)benzoate ≥95%

Supplier: Ambeed
Danger

Synonyms: Ethyl 4-(bromomethyl)benzoate

A654271-25G A654271-5G
77594-990EA 106.6 USD
77594-990 77617-740
Ethyl-4-(bromomethyl)benzoate ≥95%
Ethyl-4-(bromomethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.1 g/mol
MDL Number: MFCD03791307
CAS Number: 26496-94-6
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR