2,5,8,11,14,17,20,23,26,29,32-Undecaoxatetratriacontan-34-oic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: m-PEG10-CH2COOH

A654108-100MG A654108-250MG
77622-422EA 53.56 USD
77622-422 77622-424
2,5,8,11,14,17,20,23,26,29,32-Undecaoxatetratriacontan-34-oic acid ≥95%
2,5,8,11,14,17,20,23,26,29,32-Undecaoxatetratriacontan-34-oic acid

Precaución: for Research Use Only

Formula: C₂₃H₄₆O₁₃
MW: 530.6 g/mol
MDL Number: MFCD16619366
CAS Number: 908258-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR