Allitol ≥98%

Supplier: Ambeed
Warning

Synonyms: rel-(2R,3R,4S,5S)-Hexane-1,2,3,4,5,6-hexaol

A653889-10MG A653889-1MG A653889-5MG
77602-668EA 39.98 USD
77602-668 77602-670 77602-672
Allitol ≥98%
Allitol

Precaución: for Research Use Only

Formula: C₆H₁₄O₆
MW: 182.17 g/mol
MDL Number: MFCD11519461
CAS Number: 488-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR