Methyl 2-bromo-4-methylthiazole-5-carboxylate ≥96%

Supplier: Ambeed
Warning

A653758-1G A653758-100MG A653758-250MG
77573-096EA 99 USD
77573-096 77886-552 77886-553
Methyl 2-bromo-4-methylthiazole-5-carboxylate ≥96%
Methyl 2-bromo-4-methylthiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MDL Number: MFCD09998944
CAS Number: 81569-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR