1-Boc-4-dimethylcarbamoylpiperazine ≥95%

Supplier: Ambeed
Warning

A653304-1G A653304-250MG
77686-618EA 124 USD
77686-618 77901-378
1-Boc-4-dimethylcarbamoylpiperazine ≥95%
1-Boc-4-dimethylcarbamoylpiperazine

Precaución: for Research Use Only

Formula: C₁₂H₂₃N₃O₃
MDL Number: MFCD12406798
CAS Number: 215453-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR