1-Cyclopropyl-1H-pyrazol-4-amine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A653292-250MG A653292-100MG A653292-1G
77684-542EA 128 USD
77684-542 77891-289 77891-290
1-Cyclopropyl-1H-pyrazol-4-amine ≥98%, Ambeed.Inc
1-Cyclopropyl-1H-pyrazol-4-amine

Precaución: For research use only.

Formula: C₆H₉N₃
MDL Number: MFCD16810881
CAS Number: 1240567-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR