4-Iodo-1-butyne ≥95%

Supplier: Ambeed
Danger

A653230-1G A653230-250MG A653230-100MG A653230-5G
77573-092EA 143.17 USD
77573-092 77617-734 77749-341 77749-342
4-Iodo-1-butyne ≥95%
4-Iodo-1-butyne

Precaución: for Research Use Only

Formula: C₄H₅I
MW: 179.99 g/mol
MDL Number: MFCD20414835
CAS Number: 43001-25-8
UN: 2924
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR