7-Boc-3-hydroxy-1-oxa-7-azaspiro[4.4]nonane ≥98%

Supplier: Ambeed
Warning

A652889-100MG A652889-1G A652889-250MG A652889-5G
77893-307EA 48 USD
77893-307 77893-308 77893-309 77893-310
7-Boc-3-hydroxy-1-oxa-7-azaspiro[4.4]nonane ≥98%
7-Boc-3-hydroxy-1-oxa-7-azaspiro[4.4]nonane

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD23105883
CAS Number: 1331825-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR