2-Bromo-1-(5-chloropyridin-2-yl)ethanone ≥98%
Supplier: Ambeed
|
Danger
|
A651896-100MG
A651896-1G
A651896-5G
77884-523EA
45
USD
77884-523
77884-524
77884-525
2-Bromo-1-(5-chloropyridin-2-yl)ethanone ≥98%
2-Bromo-1-(5-chloropyridin-2-yl)ethanone
Precaución: for Research Use Only
Formula:
C₇H₅BrClNO |
MDL Number:
MFCD10697657 CAS Number: 94952-47-3 UN: 3261 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...