3-Bromo-2-methylbenzenesulfonyl chloride ≥97%
Supplier: Ambeed
|
Danger
|
A651705-250MG
A651705-5G
A651705-100MG
77884-519EA
53
USD
77884-519
77884-520
77884-518
3-Bromo-2-methylbenzenesulfonyl chloride ≥97%
3-Bromo-2-methylbenzenesulfonyl chloride
Precaución: for Research Use Only
Formula:
C₇H₆BrClO₂S |
MDL Number:
MFCD07782194 CAS Number: 886501-61-7 UN: 1759 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...