7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 7-(Trifluoromethyl)benzo[b]thiophene-2-carboxylic acid

A651356-1G A651356-5G
77673-770EA 48 USD
77673-770 77673-772
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid ≥95%
7-(Trifluoromethyl)-1-benzothiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₅F₃O₂S
MW: 246.21 g/mol
MDL Number: MFCD09027110
CAS Number: 550998-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR