(3-Fluoro-2-methoxypyridin-4-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A651308-5G A651308-250MG A651308-100MG A651308-1G
77686-613EA 768 USD
77686-613 77901-351 77901-831 77901-456
(3-Fluoro-2-methoxypyridin-4-yl)boronic acid ≥98%
(3-Fluoro-2-methoxypyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₃
MDL Number: MFCD16611247
CAS Number: 1598387-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR