(S)-2-((R)-2-Amino-4-methylpentanamido)-3-(4-hydroxyphenyl)propanoic acid hydrate ≥98%

Supplier: Ambeed
Warning

A651173-1G A651173-250MG A651173-5G
77602-664EA 65.6 USD
77602-664 77627-302 77880-368
(S)-2-((R)-2-Amino-4-methylpentanamido)-3-(4-hydroxyphenyl)propanoic acid hydrate ≥98%
(S)-2-((R)-2-Amino-4-methylpentanamido)-3-(4-hydroxyphenyl)propanoic acid hydrate
Formula: C₁₅H₂₄N₂O₅
MDL Number: MFCD00063034
CAS Number: 3303-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR