(4-(Dibenzo[b,d]furan-4-yl)phenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A651023-5G A651023-10G A651023-1G A651023-250MG
77783-584EA 234 USD
77783-584 77783-581 77783-582 77783-583
(4-(Dibenzo[b,d]furan-4-yl)phenyl)boronic acid ≥98%
(4-(Dibenzo[b,d]furan-4-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₃BO₃
CAS Number: 796071-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR