Bohemine ≥99%

Supplier: Ambeed
Warning

Synonyms: 3-((6-(benzylamino)-9-isopropyl-9H-purin-2-yl)amino)propan-1-ol

A650938-5MG A650938-100MG A650938-10MG
77686-612EA 27 USD
77686-612 77686-610 77686-611
Bohemine ≥99%
Bohemine

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₆O
MW: 340.43 g/mol
MDL Number: MFCD04974145
CAS Number: 189232-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR