Potassium cyclobutyltrifluoroborate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A650657-100MG A650657-1G A650657-250MG A650657-25G A650657-5G
77684-537EA 27 USD
77684-537 77893-301 77893-302 77893-303 77893-304
Potassium cyclobutyltrifluoroborate ≥98%, Ambeed.Inc
Potassium cyclobutyltrifluoroborate

Precaución: For research use only.

Formula: C₄H₇BF₃K
MDL Number: MFCD09992884
CAS Number: 1065010-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR