4-(((6S,9S,9aS)-1-(Benzylcarbamoyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)octahydro-2H-pyrazino[2,1-c][1,2,4]triazin-6-yl)methyl)phenyl dihydrogen phosphate 99+%

Supplier: Ambeed
Warning

Synonyms: Foscenvivint

A650610-10MG A650610-1MG A650610-5MG
77598-658EA 55.35 USD
77598-658 77598-660 77598-662
4-(((6S,9S,9aS)-1-(Benzylcarbamoyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)octahydro-2H-pyrazino[2,1-c][1,2,4]triazin-6-yl)methyl)phenyl dihydrogen phosphate 99+%
4-(((6S,9S,9aS)-1-(Benzylcarbamoyl)-2,9-dimethyl-4,7-dioxo-8-(quinolin-8-ylmethyl)octahydro-2H-pyrazino[2,1-c][1,2,4]triazin-6-yl)methyl)phenyl dihydrogen phosphate

Precaución: for Research Use Only

Formula: C₃₃H₃₅N₆O₇P
MW: 658.64 g/mol
MDL Number: MFCD12196918
CAS Number: 1422253-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR