4-Bromo-3-cyclopropyl-1H-pyrazole ≥95%

Supplier: Ambeed
Warning

A650233-1G A650233-5G
77673-768EA 49.8 USD
77673-768 77884-517
4-Bromo-3-cyclopropyl-1H-pyrazole ≥95%
4-Bromo-3-cyclopropyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MDL Number: MFCD14658056
CAS Number: 957345-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR