(E)-tert-Butyl 4-bromobut-2-enoate ≥95%

Supplier: Ambeed
Warning

A650219-1G A650219-5G
77884-515EA 310 USD
77884-515 77884-516
(E)-tert-Butyl 4-bromobut-2-enoate ≥95%
(E)-tert-Butyl 4-bromobut-2-enoate

Precaución: for Research Use Only

Formula: C₈H₁₃BrO₂
MDL Number: MFCD20484020
CAS Number: 86606-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR