AZD-3463 ≥99%

Supplier: Ambeed
Warning

Synonyms: AZD3463, N-[4-(4-Amino-1-piperidinyl)-2-methoxyphenyl]-5-chloro-4-(1H-indol-3-yl)-2-pyrimidinamine, N-(4-(4-Aminopiperidin-1-yl)-2-methoxyphenyl)-5-chloro-4-(1H-indol-3-yl)pyrimidin-2-amine

A649749-10MG A649749-5MG
77686-607EA 70 USD
77686-607 77686-608
AZD-3463 ≥99%
AZD-3463

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClN₆O
MW: 448.947 g/mol
MDL Number: MFCD25976759
CAS Number: 1356962-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR