tert-Butyl 4-amino-[1,4'-bipiperidine]-1'-carboxylate ≥95%

Supplier: Ambeed
Warning

A649591-1G A649591-100MG A649591-250MG
77614-016EA 152.44 USD
77614-016 77686-605 77686-606
tert-Butyl 4-amino-[1,4'-bipiperidine]-1'-carboxylate ≥95%
tert-Butyl 4-amino-[1,4'-bipiperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₉N₃O₂
MW: 283.40 g/mol
MDL Number: MFCD09864242
CAS Number: 959237-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR