4-(3-Aminophenyl)butanoic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

A648999-1G A648999-100MG
77627-298EA 233.81 USD
77627-298 77880-367
4-(3-Aminophenyl)butanoic acid hydrochloride ≥95%
4-(3-Aminophenyl)butanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MDL Number: MFCD04039396
CAS Number: 91843-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR