4-Formyl-2,6-dimethylphenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A648724-1G A648724-250MG
77783-576EA 126 USD
77783-576 77783-577
4-Formyl-2,6-dimethylphenylboronic acid pinacol ester ≥98%
4-Formyl-2,6-dimethylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₃
MDL Number: MFCD30344662
CAS Number: 1951423-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR