Bedaquiline fumarate ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R,2S)-1-(6-Bromo-2-methoxyquinolin-3-yl)-4-(dimethylamino)-2-(naphthalen-1-yl)-1-phenylbutan-2-ol fumarate

A648653-1MG A648653-5MG A648653-10MG
77614-004EA 6.18 USD
77614-004 77614-006 77684-532
Bedaquiline fumarate ≥98%
Bedaquiline fumarate

Precaución: For research use only.

Formula: C₃₆H₃₅BrN₂O₆
MW: 671.58 g/mol
MDL Number: MFCD28167761
CAS Number: 845533-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR