1-Bromo-8-phenyloctane ≥95%

Supplier: Ambeed
Danger

Synonyms: (8-Bromooct-1-yl)benzene, (8-Bromooctyl)benzene

A647137-1G A647137-25G A647137-5G
77669-776EA 284 USD
77669-776 77669-778 77669-780
1-Bromo-8-phenyloctane ≥95%
1-Bromo-8-phenyloctane

Precaución: for Research Use Only

Formula: C₁₄H₂₁Br
MW: 269.221 g/mol
MDL Number: MFCD01075178
CAS Number: 54646-75-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR