2-(Cyclopentyl)ethenyl-1-boronic acid ≥96% stabilised

Supplier: Ambeed
Danger

Synonyms: (E)-(2-Cyclopentylvinyl)boronic acid

Stabilization: Stabilised with TBC

A646766-250MG A646766-50MG A646766-100MG A646766-1G
77900-949EA 271 USD
77900-949 77901-212 77900-753 77900-725
2-(Cyclopentyl)ethenyl-1-boronic acid ≥96% stabilised
2-(Cyclopentyl)ethenyl-1-boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₃BO₂
MDL Number: MFCD09840294
CAS Number: 161282-93-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR