(2-Butylbenzofuran-3-yl)(4-(2-(ethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride ≥98%

Supplier: Ambeed
Warning

A645883-100MG A645883-250MG A645883-50MG
77686-600EA 399 USD
77686-600 77901-678 77900-735
(2-Butylbenzofuran-3-yl)(4-(2-(ethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride ≥98%
(2-Butylbenzofuran-3-yl)(4-(2-(ethylamino)ethoxy)-3,5-diiodophenyl)methanone hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₆ClI₂NO₃
MDL Number: MFCD08060094
CAS Number: 96027-74-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR