1-(3-Chloro-5-((4-(4-chlorothiophen-2-yl)-5-(4-cyclohexylpiperazin-1-yl)thiazol-2-yl)carbamoyl)pyridin-2-yl)piperidine-4-carboxylic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Avatrombopag
A645681-1G
A645681-100MG
A645681-50MG
77614-002EA
47.38
USD
77614-002
77684-527
77684-528
1-(3-Chloro-5-((4-(4-chlorothiophen-2-yl)-5-(4-cyclohexylpiperazin-1-yl)thiazol-2-yl)carbamoyl)pyridin-2-yl)piperidine-4-carboxylic acid ≥98%
1-(3-Chloro-5-((4-(4-chlorothiophen-2-yl)-5-(4-cyclohexylpiperazin-1-yl)thiazol-2-yl)carbamoyl)pyridin-2-yl)piperidine-4-carboxylic acid
Precaución: For research use only.
Formula:
C₂₉H₃₄Cl₂N₆O₃S₂ MW: 649.65 g/mol |
MDL Number:
MFCD18633249 CAS Number: 570406-98-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...