2-Carbomethoxybenzylamine hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-(aminomethyl)benzoate hydrochloride

A645455-100MG A645455-1G A645455-250MG
77878-457EA 20 USD
77878-457 77878-458 77878-459
2-Carbomethoxybenzylamine hydrochloride ≥95%
2-Carbomethoxybenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClNO₂
MDL Number: MFCD06245496
CAS Number: 849020-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR