Iminodiacetic acid disodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Sodium 2,2'-azanediyldiacetate, Iminodiacetic acid disodium salt, Disodium Iminodiacetate hydrate

A644767-500G A644767-100G A644767-25G
77669-768EA 84 USD
77669-768 77880-363 77880-364
Iminodiacetic acid disodium salt ≥98%
Iminodiacetic acid disodium salt

Precaución: for Research Use Only

Formula: C₄H₅NNa₂O₄
MDL Number: MFCD00043340
CAS Number: 928-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR