Methyl 2-((4-hydroxyphenyl)amino)acetate ≥95%

Supplier: Ambeed
Warning

A644545-1G A644545-250MG
77884-506EA 36 USD
77884-506 77884-507
Methyl 2-((4-hydroxyphenyl)amino)acetate ≥95%
Methyl 2-((4-hydroxyphenyl)amino)acetate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MDL Number: MFCD08063907
CAS Number: 56405-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR