5-Methyl-1H-indazole-6-boronic acid ≥97%

Supplier: Ambeed
Warning

A644262-100MG A644262-1G A644262-250MG
77686-597EA 16 USD
77686-597 77901-237 77901-346
5-Methyl-1H-indazole-6-boronic acid ≥97%
5-Methyl-1H-indazole-6-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD12028648
CAS Number: 1310383-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR