2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Methoxy-5-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A644093-100MG A644093-250MG A644093-1G A644093-5G
77749-324EA 31 USD
77749-324 77749-326 77749-325 77749-327
2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester ≥95%
2-Methoxy-5-trifluoromethoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BF₃O₄
MW: 318.097 g/mol
MDL Number: MFCD22415050
CAS Number: 1688666-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR