2-Ethoxypyridine-4-boronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A643648-250MG A643648-1G A643648-5G
77669-762EA 53.71 USD
77669-762 77873-040 77873-041
2-Ethoxypyridine-4-boronic acid pinacol ester ≥98%
2-Ethoxypyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₃
MDL Number: MFCD11617874
CAS Number: 957346-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR