Phenanthren-3-amine ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
phenanthren-3-amine
A643473-1G
A643473-250MG
A643473-100MG
77886-534EA
153
USD
77886-534
77886-535
77886-533
Phenanthren-3-amine ≥98%
Phenanthren-3-amine
Precaución: for Research Use Only
Formula:
C₁₄H₁₁N MW: 193.24 g/mol |
MDL Number:
MFCD00019108 CAS Number: 1892-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...